Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Singapore
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Sodium thiosulphate pentahydrate
Sodium thiosulphate pentahydrate AR, Rankem®
Sodium thiosulphate pentahydrate AR, Rankem®
Sodium thiosulphate pentahydrate AR, Rankem®
Catalog # RANKS0439
Supplier:
RANKEM
CAS Number:
Certificates
Expand image
Sodium thiosulphate pentahydrate AR, Rankem®
Catalog # RANKS0439
Supplier:
RANKEM
CAS Number:
Certificates
Specifications
Pk:
1 kg
Seq:
4024
Specifications
About this item
Product Details
Documentation
:
:
248.19 g/mol
CAS Number:
10102-17-7
Formula:
Na₂S₂O₃.5H₂O
MDL Number:
MFCD00149186
Product Details
Documentation