
Sodium hydroxide ≥97% LR, Laboratory Reagent, Rankem®
Supplier: RANKEM
Certificates
About this item
CAS Number:1310-73-2
:
Formula:NaOH
MDL Number:MFCD00003548
:40 g/mol
Boiling Pt:1390 °C (1013 hPa)
Melting Pt:323 °C
Density:2,13 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:215-185-5
UN:1823
ADR:8,II
:GHS05
Specification Test Results
- Guarantee Analysis:.
- Assay:Min. 97.00 %
- Identity:Passes test
- Maximum Limits Of Impurities:.
- Carbonate(Na2CO3):Max. 1.50 %
- Chloride (Cl):<=0.01%
- Heavy Metals (As Pb):<=0.001%
- Iron (Fe):<=0.005%
- Sulphate (SO4):<=0.01%