Order Entry
Singapore
ContactUsLinkComponent
2-Propanol ≥99.0% LR, Laboratory Reagent, Rankem®
2-Propanol ≥99.0% LR, Laboratory Reagent, Rankem®
Supplier:  RANKEM
Certificates
undefined
2-Propanol ≥99.0% LR, Laboratory Reagent, Rankem®
Supplier:  RANKEM
Certificates

Specification Test Results

  • Guarantee Analysis:
    .
  • Assay(GC):
    Min. 99.00 %
  • Weight Per ml (at 20°C):
    0.784 --> 0.786 g
  • Identity (IR):
    Conforms
  • Maximum Limits Of Impurities:
    .
  • Residue After Evaporation:
    Max. 0.002 %
  • Water:
    Max. 0.20 %

Specification Test Results