Order Entry
Singapore
ContactUsLinkComponent
tert-Butyl methyl ether AR, Rankem®
tert-Butyl methyl ether AR, Rankem®
Catalog # RANKM2515
Supplier:  RANKEM
CAS Number:  
undefined
tert-Butyl methyl ether AR, Rankem®
Catalog # RANKM2515
Supplier:  RANKEM
CAS Number:  

Specifications

  • Pk:
    2,5 L
  • Seq:
    5096

Specifications

About this item

 :
 :88.15 g/mol
Storage Temperature:Ambient
UN:2398
ADR:3,II
 :GHS02
 :GHS07
 :GHS08
CAS Number:1634-04-4
Formula:(CH₃)₃COCH₃
MDL Number:MFCD00008812