Methanol ≥99.00%, Laboratory Reagent, Rankem®
: RANKEM
:67-56-1
:
:CH₃OH
:MFCD00004595
:32.04 g/mol
:–98 °C
:0,7918 g/cm³ (20 °C)
:11 °C (closed cup)
:1230
:GHS02
:GHS06
:GHS08
:Ambient
- Assay (GC):Min. 99.00%
- Weight Per ml (at 20 °C):0.791 - 0.793 g
- Refractive Index (at 20 °C):1.328 - 1.331
- Distillation Range:64.0 - 65.0 °C
- Non-Volatile Matter:Max. 0.0050%
- Aldehydes And Ketones:≤0.01%
- Water ( By KF):Max. 0.2000%
- Readily Carbonisable Substances:Passes Test