

Acetone ≥99.70%, Laboratory Reagent, Rankem®
: RANKEM
:67-64-1
:
:CH₃COCH₃
:MFCD00008765
:58.08 g/mol
:56,2 °C (1013 hPa)
:–95,4 °C
:0,792 g/cm³ (20 °C)
:<–20 °C (closed cup)
:Ambient
:200-662-2
:1090
:3,II
:GHS02
:GHS07
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental certification:Chem21 Solvent Selection Guide recommended after discussion
- Purity:≥ 99.70%
- Relative Density at 27/27 °C:0.784 - 0.786
- Color:≤ 10 APHA
- Residue On Evaporation, mg/100 ml:≤ 5.00
- Water content:≤ 0.4%
- Acidity (As CH₃COOH) g/100 ml:≤ 0.002
- Alkalinity:Passes test
- Permanganate Test:≥ 30 min
- Alcoholic Impurities:Passes test
- Benzene content:≤ 2.00 mg/kg
- Methanol content:≤ 300.0 mg/kg
- Identity (IR):Conforms
- Weight Per ml (at 20°C):0.789 - 0.791 g