

Acetonitrile ≥99.80% (by GC) for HPLC, Rankem®
Supplier: RANKEM
Certificates
About this item
CAS Number:75-05-8
:
Formula:H₃CCN
MDL Number:MFCD00001878
:41.05 g/mol
UN:1648
ADR:3,II
:GHS02
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental certification:EPA Safer Chemical Solvent (Green Circle Rating); Chem21 Solvent Selection Guide recommended by default and after discussion
Specifications
Specification Test Results
- Assay (GC):Min. 99.80%
- Weight per ml (at 20 °C):0.782 - 0.783 g
- Acidity:Acidity Max. 0.000200 meq/g
- Alkalinity:Max. 0.0002 meq/g
- Non-Volatile Matter:Max. 0.00050%
- Water ( By KF):Max. 0.0500%
- UV Transmittance Wavelength, 195 nm:Min. 70.00%
- UV Transmittance Wavelength, 200 nm:Min. 90.00%
- UV Transmittance Wavelength, From 240 nm:Min. 98.00%