Order Entry
Singapore
ContactUsLinkComponent
Cyclohexane ≥99.8%, BAKER ANALYZED®, J.T.Baker®
Cyclohexane ≥99.8%, BAKER ANALYZED®, J.T.Baker®
Supplier:  Avantor
Certificates
undefined
Cyclohexane ≥99.8%, BAKER ANALYZED®, J.T.Baker®
Supplier:  Avantor
Certificates

Specification Test Results

  • Assay:
    Min. 99.8%
  • Appearance:
    Clear
  • Colour:
    Max. 10 ppb APHA
  • Water:
    Max. 0.01%
  • Acidity:
    Max. 0.0003 mEq/g
  • Residue after Evaporation:
    Max. 0.001%
  • Cyclohexane:
    Max. 0.05%
  • Ethanol:
    Max. 0.01%
  • Chloride:
    Max. 0.2 ppm
  • Phospahte:
    Max. 0.5 ppm
  • Aluminium:
    Max. 50 ppb
  • Arsenic:
    Max. 20 ppb
  • Barium:
    Max. 10 ppb
  • Boron:
    Max. 10 ppb
  • Cadmium:
    Max. 20 ppb
  • Calcium:
    Max. 50 ppb
  • Chromium:
    Max. 10 ppb
  • Cobalt:
    Max. 10 ppb
  • Copper:
    Max. 10 ppb
  • Gallium:
    Max. 20 ppb
  • Germanium:
    Max. 20 ppb
  • Gold:
    Max. 20 ppb
  • Iron:
    Max. 50 ppb
  • Lead:
    Max. 20 ppb
  • Lithium:
    Max. 20 ppb
  • Magnesium:
    Max. 20 ppb
  • Manganese:
    Max. 20 ppb
  • Nickiel:
    Max. 20 ppb
  • Potassium:
    Max. 50 ppb
  • Silicon:
    Max. 20 ppb
  • Silver:
    Max. 20 ppb
  • Sodium:
    Max. 50 ppb
  • Strontium:
    Max. 20 ppb
  • Tin:
    Max. 20 ppb
  • Zinc:
    Max. 50 ppb

Specification Test Results

Related Information

Certificates
Search by Lot Number