Order Entry
Singapore
ContactUsLinkComponent
 
Flammable
Irritant
Dimethyldichlorosilane silylation reagent, for GC derivatisation
Catalog # MANA701230.650
Supplier:  MACHEREY-NAGEL
CAS Number:  
undefined
Flammable
Irritant
Dimethyldichlorosilane silylation reagent, for GC derivatisation
Catalog # MANA701230.650
Supplier:  MACHEREY-NAGEL
CAS Number:  

Specifications

  • Pack type:
    Glass vial, amber, screw cap
  • Pk:
    1 KIT
  • Seq:
    0075
  • Pk Info:
    6× 50 ml

Specifications

About this item

 :
 :129.06 g/mol
Boiling Pt:70 °C (760 mm Hg)
Density:1,07 g/cm³ (d20°/4°)
UN:3316
ADR:9,II
 :GHS02
 :GHS07
CAS Number:75-78-5
Formula:C₂H₆Cl₂Si
MDL Number:MFCD00000491