


1-(Trimethylsilyl)imidazole in pyridine Silyl-1139 silylation reagent, for GC derivatisation
Catalog # MANA701460.201
Supplier: MACHEREY-NAGEL
CAS Number:
Specifications
- Pack type:Glass vial, amber, crimped
- Pk:1 KIT
- Seq:0075
- Pk Info:20× 1 ml
Specifications
About this item
:
:140.26 g/mol
Flash Pt:17 °C
Storage Temperature:Refrigerator
UN:3316
ADR:9,II
:GHS02
:GHS07
:GHS08
CAS Number:18156-74-6
Formula:C₆H₁₂N₂Si
MDL Number:MFCD00005280