Pharmaceutical analytical standard, 2-NP-SCA-13C,15N2, reference material, Honeywell Fluka™
HONCR1660-10MG
:
- Pack type:Glass Bottle
- Pk:10 mg
- Standards:Analytical Standard
- CAS No.:957509-32-9
- Description:2-NP-SCA-¹³C,¹⁵N2
- Formula:C₇13CH₈N₂15N₂O₃
- Storage temperature:Ambient
- Description:2-NP-SCA-¹³C,¹⁵N2
- Form:Crystalline
High quality organic reference materials produced according to ISO 9001.
: Supplied with complete Certificate of Analysis (CoA) with expiry date and lot specific analytical results.