Pharmaceutical analytical standard, 2-NP-AMOZ, reference material, Honeywell Fluka™
Catalog # HONCR1656-10MG
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Glass Bottle
- Pk:10 mg
- Standards:Analytical Standard
- CAS No.:183193-59-1
- Description:2-NP-AMOZ
- Formula:C₁₅H₁₈N₄O₅
- Storage temperature:Ambient
- Description:2-NP-AMOZ
- Form:Crystalline
Specifications
About this item
High quality organic reference materials produced according to ISO 9001.
Delivery: Supplied with complete Certificate of Analysis (CoA) with expiry date and lot specific analytical results.