Pharmaceutical analytical standard, Tilmicosin, reference material, Honeywell Fluka™
Catalog # HONCR1646-100MG
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Glass Bottle
- Pk:100 mg
- Standards:Analytical Standard
- CAS No.:108050-54-0
- Description:Tilmicosin
- Formula:C₄₆H₈₀N₂O₁₃
- Storage temperature:Ambient
- Description:Tilmicosin
- Form:Solid
Specifications
About this item
High quality organic reference materials produced according to ISO 9001.
Delivery: Supplied with complete Certificate of Analysis (CoA) with expiry date and lot specific analytical results.