Organic analytical standards, Toluene, reference material, Honeywell Fluka™
HONCR1592-5ML
:
- Pack type:Glass Bottle
- Pk:5 ml
- Standards:Analytical Standard
- CAS No.:108-88-3
- Description:Toluene
- Formula:C₆H₅CH₃
- Storage temperature:Ambient
- Description:Toluene
- Form:Liquid
High quality organic reference materials produced according to ISO 9001.
: Supplied with complete Certificate of Analysis (CoA) with expiry date and lot specific analytical results.