



Iodine monochloride solution, StandARd®, Macron Fine Chemicals™
Catalog # MACRH180-05
Supplier: MACRON AVANTOR BRAND
CAS Number:
Specifications
- Pack type:Glass bottle, amber, narrow mouth
- Pk:1 L
- Seq:5037
Specifications
Specification Test Results
- Assay (halogen content as Normality) (at time of packaging):0.215 - 0.225
Specification Test Results
About this item
:
:162.36 g/mol
UN:2920
ADR:8,3,II
:GHS02
:GHS05
:GHS08
:GHS07
CAS Number:7790-99-0
Formula:ClI
MDL Number:MFCD00011354