Order Entry
Singapore
ContactUsLinkComponent
Test papers, qualitative, ammonium ions and ammonia
Test papers, qualitative, ammonium ions and ammonia
Catalog # MANA90722
Supplier:  MACHEREY-NAGEL
CAS Number:  
undefined
Test papers, qualitative, ammonium ions and ammonia
Catalog # MANA90722
Supplier:  MACHEREY-NAGEL
CAS Number:  

Specifications

  • Chemical method:
    Nesslers reagent
  • Pk:
    200
  • Description:
    Ammonia and ammonium ions (NH₃, NH₄⁺)
  • Range:
    >10 mg/l NH₄⁺

Specifications

About this item

The test paper can be used for the rapid detection of Ammonium ions as well as for gaseous Ammonia. The white paper turns brownish-yellow in the presence of Ammonium ions or NH₃.

  • Specific for the determination of NH₄⁺

Colour changes: White to brown-yellow