Order Entry
Singapore
ContactUsLinkComponent
Test strips, potassium
Test strips, potassium
Catalog # MANA90727
Supplier:  MACHEREY-NAGEL
CAS Number:  
undefined
Test strips, potassium
Catalog # MANA90727
Supplier:  MACHEREY-NAGEL
CAS Number:  

Specifications

  • Chemical method:
    Dipicrylamine
  • Pk:
    200
  • Description:
    Potassium ions
  • Range:
    250 mg/l K⁺

Specifications

About this item

The test paper allows the quick and easy detection of Potassium. Rubidium, Cesium and Thallium(I) give the same reaction. Sodium and heavy metal ions decrease the sensitivity.

  • Colour changes: Bright-yellow to orange

The leaflet describes a method to eliminate these interferences.