Order Entry
Singapore
Orders LinkContactUsLinkComponent
QUANTOFIX® Copper Sensitive 5

QUANTOFIX® Copper Sensitive 5

Test paper phosphatesmo MI for phosphatase in milk, 0,5% raw milk in pasteurised milk; 300 U/L alkaline phosphatase in pasteurised milk, Color reaction with an organic phosphate in alkaline pH range, 40×40×120 mm (packaging), 50 Tests, 50
Catalog # MANA90612
Supplier:   MACHEREY-NAGEL
QUANTOFIX® Copper Sensitive 5
Catalog # MANA90612
Supplier:  MACHEREY-NAGEL
CAS Number:  
QUANTOFIX® Copper Sensitive 5
QUANTOFIX® Copper Sensitive 5

Test paper phosphatesmo MI for phosphatase in milk, 0,5% raw milk in pasteurised milk; 300 U/L alkaline phosphatase in pasteurised milk, Color reaction with an organic phosphate in alkaline pH range, 40×40×120 mm (packaging), 50 Tests, 50

CAS Number:  
Price
Inquire For Price
Log in for availability
1 * 50 items
Product Details & Documents

Semi-quantitative test strips for reliable determination of copper concentrations as low as 0,1 mg/L (ppm).

  • Sensitive rapid test

A hole in the plastic carrier allows the water to migrate through the test pad increasing the sensitivity significantly. Ideal for rapid and easy analysis of copper directly on-site. No maintenance or calibration is required.

Specifications

Specifications

  • Chemical method:
    Color reaction with an organic phosphate in alkaline pH range
  • Pk:
    50
  • Description:
    Test paper phosphatesmo MI for phosphatase in milk
  • Dimensions:
    40×40×120 mm (packaging)
  • Tests per kit:
    50 Tests
  • Range:
    0,5% raw milk in pasteurised milk; 300 U/L alkaline phosphatase in pasteurised milk
  • Cat. No.:
    MANA90612
  • Contents:
    50 test strips in a tube
  • Platform:
    Qualitative test paper
  • Compliance:
    Not CE certified and no hazardous material
  • Shelf life (months):
    12 (from production)
  • W×D×H:
    10×85 mm (net)
  • Method:
    Phosphatase - white → yellow
  • Weight:
    46 g

Specifications