Organic analytical standards, beta-Zearalenol-Zearalenol-d5, neat, reference material, Honeywell Fluka™
Catalog # HONCR1438-10MG
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:10 mg
- Description:beta-Zearalenol-Zearalenol-d₅
- Formula:C₁₈H₁₉D₅O₅
- Storage temperature:–20 °C
- Description:beta-Zearalenol-Zearalenol-d₅
- Freezing point:157...159 °C
Specifications
About this item
- Ensure you have the information you need to quickly start your analysis, and pass relevant audits
- High quality inorganic and organic reference materials produced according to ISO 9001
- Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results