Pesticide reference standard, Pyrazoxyfen, neat, Honeywell Fluka™
Catalog # HONCR1130-50MG
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:50 mg
- CAS No.:71561-11-0
- Description:Pyrazoxyfen
- Formula:C₂₀H₁₆Cl₂N₂O₃
- Storage temperature:Ambient
- Description:Pyrazoxyfen
Specifications
About this item
High quality inorganic and organic reference materials produced according to ISO 9001.
- Ensure you have the information you need to quickly start your analysis, and pass relevant audits
- Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results