Organic analytical standards, [D-Asp3, E-Dhb7]-Microcystin-RR solution, 10 μg/ml in methanol, reference material, Honeywell Fluka™
Catalog # HONCR1095-1ML
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Concentration:10 μg/ml in methanol
- Pack type:Glass bottle
- Pk:1 mL
- CAS No.:202120-08-9
- Description:[D-Asp3, E-Dhb7]-Microcystin-RR solution
- Formula:C₄₈H₇₃N₁₃O₁₂
- Storage temperature:–20 °C
- Description:[D-Asp3, E-Dhb7]-Microcystin-RR solution
- Flash point:11 °C
- Boiling point:64...65 °C
- Freezing point:–98 °C
Specifications
About this item
High quality inorganic and organic reference materials produced according to ISO 9001.
- Ensure you have the information you need to quickly start your analysis, and pass relevant audits
- Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results