Order Entry
Singapore
ContactUsLinkComponent
Pesticide reference standard, 1,2,3,4-Tetrachloro-5-nitrobenzene, neat, Honeywell Fluka™
Pesticide reference standard, 1,2,3,4-Tetrachloro-5-nitrobenzene, neat, Honeywell Fluka™
Catalog # HONCR1076-100MG
CAS Number:  
Pesticide reference standard, 1,2,3,4-Tetrachloro-5-nitrobenzene, neat, Honeywell Fluka™
Catalog # HONCR1076-100MG
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 mg
  • CAS No.:
    879-39-0
  • Description:
    1,2,3,4-Tetrachloro-5-nitrobenzene
  • Formula:
    C₆HCl₄NO₂
  • Storage temperature:
    Ambient
  • Description:
    1,2,3,4-Tetrachloro-5-nitrobenzene
  • Freezing point:
    65...67 °C

Specifications

About this item

High quality inorganic and organic reference materials produced according to ISO 9001.

  • Ensure you have the information you need to quickly start your analysis, and pass relevant audits
  • Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results