VISOCOLOR® Colorimetric Test, Chloride
Supplier: MACHEREY-NAGEL
Certificates
About this item
Reagents for the determination of chloride in water samples.
- Combines simple implementation with high measurement reliability
Simple determination directly on site or in the laboratory.
Specifications
- Model:MANA931018
- Platform:Colorimetric test kits
- Storage temperature:15...25 °C
- Shelf life (months):12
- Weight:252 g (Including packaging)