Order Entry
Singapore
ContactUsLinkComponent
Test papers, qualitative, aluminum/zirconium
Test papers, qualitative, aluminum/zirconium
Catalog # MANA90721
Supplier:  MACHEREY-NAGEL
CAS Number:  
undefined
Test papers, qualitative, aluminum/zirconium
Catalog # MANA90721
Supplier:  MACHEREY-NAGEL
CAS Number:  

Specifications

  • Chemical method:
    Alizarin red
  • Pk:
    100
  • Description:
    Aluminium/Zirconium test papers

Specifications

About this item

The test paper allows the quick and easy detection of aluminium. In the presence of aluminium (Al3+) the paper shows bright red spots on a yellow background.

  • Using a different procedure, the test paper can be used for the determination of zirconium

Colour change: Yellow to bright-red (aluminium)/red to violet (zirconium)

Limit of sensitivity: 10 mg/L Al3+

Delivery: Pack of 100 strips 20×70 mm.