

Petroleum spirit, 30…75 °C, AR®, Macron Fine Chemicals™
MACR4976-08
:
- Pack type:Glass bottle, amber, narrow mouth
- Pk:4 L
- Seq:5162
- For Laboratory,Research,or Manufacturing Use:Passes Test
- Acidity:Passes Test
- Appearance and Color:Passes Test
- Residue after Evaporation:≤0.0010 %
- Sulfur Compounds (doctor test):Passes Test
- Boiling Range(Lower):30.0 - 75.0 °C
- Boiling Range(Upper):30.0 - 75.0 °C
:
:1268
:3,II
:GHS02
:GHS08
:64742-49-0
:64742-82-1
:MFCD00081849