
Lactic acid 85%, AR® ACS, Macron Fine Chemicals™
Catalog # MACR2676-04
Supplier: MACRON AVANTOR BRAND
CAS Number:
Specifications
- Pack type:PE bottle
- Pk:500 g
- Seq:3645
Specifications
Specification Test Results
- ACS - Assay (CH₃CHOHCOOH):85.0 - 90.0 %
- ACS - Chloride (Cl):≤ 0.001 %
- ACS - Heavy Metals (by ICP-OES):≤ 5 ppm
- ACS - Iron (Fe):≤ 5 ppm
- ACS - Residue after Ignition:≤ 0.02 %
- ACS - Substances Darkened by H₂SO₄:Passes Test
- ACS - Sulfate (SO₄):≤ 0.002 %
Specification Test Results
About this item
:
Density:1,20 g/cm³
Flash Pt:112,00 °C
UN:3265
ADR:8,II
:Management of Change (MOC) category = N
:90.08 g/mol
:GHS05
CAS Number:50-21-5
:598-82-3
Formula:H₃CCH(OH)COOH
MDL Number:MFCD00004520