Order Entry
Singapore
ContactUsLinkComponent
Triethylammonium phosphate solution ~1.0 M in water, BioUltra, Fluka™
Triethylammonium phosphate solution ~1.0 M in water, BioUltra, Fluka™
Catalog # HONC90361-100ML
CAS Number:  
Triethylammonium phosphate solution ~1.0 M in water, BioUltra, Fluka™
Catalog # HONC90361-100ML
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 mL
  • Seq:
    2440

Specifications

Specification Test Results

  • Assay:
    0.9 - 1.1 mol/l
  • Refractive index (n 20/D):
    1.358 - 1.362
  • pH (direct, 25 °C):
    2.9 - 3.1
  • Arsenic (As):
    Max. 0.1 ppm
  • Aluminium (Al):
    Max. 1 ppm
  • Barium (Ba):
    Max. 1 ppm
  • Bismuth (Bi):
    Max. 1 ppm
  • Calcium (Ca):
    Max. 5 ppm
  • Cadmium (Cd):
    Max. 1 ppm
  • Cobalt (Co):
    Max. 1 ppm
  • Chromium (Cr):
    Max. 1 ppm
  • Copper (Cu):
    Max. 1 ppm
  • Iron (Fe):
    Max. 1 ppm
  • Potassium (K):
    Max. 20 ppm
  • Lithium (Li):
    Max. 1 ppm
  • Magnesium (Mg):
    Max. 1 ppm
  • Manganese (Mn):
    Max. 1 ppm
  • Molybdenum (Mo):
    Max. 1 ppm
  • Sodium (Na):
    Max. 20 ppm
  • Nickel (Ni):
    Max. 1 ppm
  • Lead (Pb):
    Max. 1 ppm
  • Strontium (Sr):
    Max. 1 ppm
  • Zinc (Zn):
    Max. 1 ppm
  • Sulfate (SO₄):
    Max. 10 ppm
  • Chloride (Cl):
    Max. 10 ppm
  • Filter test:
    complying
  • Absorbance 280 nm:
    Max. 0.03
  • Absorbance at 260 nm:
    Max. 0.04

Specification Test Results

About this item

 :
 :199.19 g/mol
Density:1,050 g/cm³
Storage Temperature:Ambient
CAS Number:10138-93-9
Formula:C₆H₁₈NO₄P
MDL Number:MFCD00067475