Order Entry
Singapore
ContactUsLinkComponent
 
Irritant
Health
Flammable
Toluene ≥99.8%, B&J Brand™ for HPLC, for gas chromatography, for spectrophotometry, Burdick & Jackson™
Catalog # BJAA347-4
Supplier:  BURDICK JACKSON
CAS Number:  
undefined
Irritant
Health
Flammable
Toluene ≥99.8%, B&J Brand™ for HPLC, for gas chromatography, for spectrophotometry, Burdick & Jackson™
Catalog # BJAA347-4
Supplier:  BURDICK JACKSON
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    4 L
  • Seq:
    5162

Specifications

Specification Test Results

  • Benzene:
    500 mg/L max.
  • Electron Capture GC:
    2 µg/L max.
  • GC Analysis:
    99.8 % min.
  • Refractive Index (20ºC):
    1.4963 - 1.4971
  • Residue:
    1 mg/L max.
  • UV Absorbance @ 300 nm:
    0.12 AU max.
  • UV Absorbance @ 325 nm:
    0.025 AU max.
  • UV Cutoff:
    284 nm max.
  • Water by Karl Fischer Titration:
    0.03 % max.

Specification Test Results

About this item

 :
 :92.14 g/mol
Boiling Pt:110,6 °C
Melting Pt:–95 °C
Density:0,867 g/cm³ (20 °C)
Flash Pt:7,1 %(V)
UN:1294
ADR:3,II
 :GHS07
 :GHS08
 :GHS02
Storage Temperature:Ambient
CAS Number:108-88-3
Formula:C₆H₅CH₃
MDL Number:MFCD00008512