Semi automated Clinical Chemistry Analyser BeneSphera™ C-61
BENP20001751
- Description:Semi automated Clinical chemistry Analyser BeneSphera™ C-61
- Environmentally Preferable:
- Pk:1
BeneSphera™ C-61 semi-automated clinical chemistry analyser offers high precision and excellent reproducibility for reliable reporting.
- Excellent QC function with the option of 2 levels of programmable controls per test
- Double aspiration air gap after each sample prevents cross-contamination
- Large LCD display allows for easy keypad operation
- Memory for 60 programs and 2000 test results - suitable for running profiles