Reagent case for fish analysis, VISOCOLOR®
Supplier: MACHEREY-NAGEL
Certificates
About this item
The VISOCOLOR® fish analysis case was specially developed for the examination of fishing waters. The rugged case with a premium foam inlay is particularly suitable for mobile analysis directly at the sampling place.
- Compact mini-lab for fishing waters
- Determination of the most important water parameters
- Colour-coded reagents
- High flexibility and incredible price
Besides the equipment, which is necessary for the analysis, the VISOCOLOR® fish case includes 6 tests for the colorimetric and titrimetric determination of the most important parameters for fishing waters.
Benefits include maximum safety due to exact labeling of all reagents, suitable for amateurs, reliable results due to color and turbidity compensation, stable and rugged case as well as chemical resistant foam inlay and safe for the environment and easy waste disposal of the used tests.
Delivery: VISOCOLOR® fish reagent case including the necessary reagents and equipment for determination of ammonium, total hardness, nitrate, nitrite, phosphate and pH, the manual and colour chart.