Organic reference standard, oxybutynin hydrochloride
Catalog # EDQMO0288000
Supplier: EDQM
CAS Number:
Specifications
- Pk:150 mg
- Standards:Reference standard
- CAS No.:1508-65-2
- Description:Oxybutynin hydrochloride
- Formula:C₂₂H₃₂ClNO₃
- Storage temperature:5 ±3 °C
- Description:Oxybutynin hydrochloride
Specifications
About this item
- Filled under inert atmosphere protected from humidity