


5-(Ethylthio)-1H-tetrazole (ETT) 0.3M in acetonitrile + 0.5% NMI, BioSyn™ activator reagent, for DNA/RNA synthesis, Burdick & Jackson™
Catalog # BJAABR731-0450
Supplier: BURDICK JACKSON
CAS Number:
Specifications
- Pk:5.400 ml
- Seq:2925
- Pk Info:12×450 ml
Specifications
About this item
:
Boiling Pt:82 °C
Density:0,8028 g/cm3
Flash Pt:11 °C
ADR:3,II
:130.17 g/mol
UN:1648
:GHS02
:GHS08
:GHS07
Storage Temperature:Ambient
CAS Number:89797-68-2
Formula:C₃H₆N₄S
MDL Number:MFCD00068733