Order Entry
Singapore
ContactUsLinkComponent
 
 
 
Formic acid : Triethylamine concentrate (1:1) 2 M:2 M in water for HPLC, Fluka™
  HONC09752-100ML
 :  
 
 
Formic acid : Triethylamine concentrate (1:1) 2 M:2 M in water for HPLC, Fluka™
  HONC09752-100ML
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    100 mL
  • Seq:
    2440

 

 

  • Density (D 20/4):
    1.005 - 1.014
  • Refractive index (n 20/D):
    1.354 - 1.374
  • Suitability test (HPLC):
    complying
  • Absorbance 500 nm:
    Max. 0.01
  • Absorbance at 290 nm:
    Max. 0.02
  • Absorbance 280 nm:
    Max. 0.03
  • Absorbance at 270 nm:
    Max. 0.03
  • Absorbance at 260 nm:
    Max. 0.1

 

 

 :
 :89 °C
 :1,01 g/cm³ (20 °C)
 :Ambient
 :1760
 :8,II
 :GHS07
 :GHS05
 :64-18-6
 :MFCD00134943