Order Entry
Singapore
ContactUsLinkComponent
 
Talc, powder, meets analytical specification of BP, Ph. Eur., Fluka™
  HONC18654-25KG
 :  
Talc, powder, meets analytical specification of BP, Ph. Eur., Fluka™
  HONC18654-25KG
 :  

 

  • Pack type:
    Plastic bottle
  • Pk:
    25 kg
  • Seq:
    4283
  • Environmentally Preferable:

 

 

  • Loss on ignition (1000°C):
    Max. 7.0 %
  • Water soluble matter:
    Max. 0.2 %
  • Acidity or alkalinity:
    complying
  • Aluminium (Al):
    Max. 2.0 %
  • Calcium (Ca):
    Max. 0.9 %
  • Iron (Fe):
    Max. 0.25 %
  • Magnesium (Mg):
    17.0 - 19.5 %
  • Lead (Pb):
    Max. 10 ppm
  • Microbiological impurity:
    complying
  • Identity:
    complying
  • Residual solvents:
    complying

 

 

 : Magnesium silicate monohydrate, Soapstone, Talcum venetum
 :
 :379.27 g/mol
 :800 °C
 :2.7 g/cm3 (20 °C)
 :Ambient
 :238-877-9
 :14807-96-6
 :Mg₃O₁₁Si₄·1H₂O
 :MFCD00792903
 :13,09127