Bromocresol green Reag. Ph. Eur., Fluka™
Supplier: Honeywell Chemicals
Certificates
About this item
CAS Number:76-60-8
:
Formula:C₂₁H₁₄Br₄O₅S
MDL Number:MFCD00005874
:698.02 g/mol
Merck Index:13,01370
Melting Pt:225 °C
Storage Temperature:Ambient
Specification Test Results
- pH Range:3.8 - 5.4, yellow to blue
- Indicator solution:complying
- Change range:complying
- Test of sensitivity:complying
- Curve course (UV):complying