
Potassium hexacyanoferrate(III) ≥99%, Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Fluka™
Supplier: Honeywell Chemicals
Certificates
About this item
CAS Number:13746-66-2
:
Formula:K₃Fe(CN)₆
MDL Number:MFCD00011392
:329.25 g/mol
Merck Index:13,07712
Boiling Pt: Dec. (on heating)
Melting Pt: Dec.
Density:1,85 g/cm³ (20 °C)
EINECS:237-323-3
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Assay:Min. 99 %
- Water insoluble matter:Max. 0.005 %
- Calcium (Ca):Max. 0.0005 %
- Cadmium (Cd):Max. 0.0005 %
- Cobalt (Co):Max. 0.005 %
- Copper (Cu):Max. 0.001 %
- Sodium (Na):Max. 0.02 %
- Nickel (Ni):Max. 0.001 %
- Lead (Pb):Max. 0.002 %
- Zinc (Zn):Max. 0.0005 %
- Chloride (Cl):Max. 0.005 %
- Hexacyanoferrate(II) ([Fe(CN)₆]₄⁻):Max. 0.02 %
- Sulfate (SO₄):Max. 0.005 %