

Diethyl ether ≥99.8% stabilised, CHROMASOLV™ for pesticide residue analysis, Riedel-de Haën™
HONC31671-6X1L
:
- Pack type:Glass bottle
- Pk:6 L
- Seq:5198
- Assay GC (Product + Ethanol):Min. 99.8 %
- Refractive Index:n20/D 1.353
- Non-volatile matter:Max. 0.0005 %
- Water (Karl Fischer):Max. 0.05 %
- Autoignition Temperature:320 °F
- Refractive index (n 20/D):1.352 - 1.354
- Suitability for residue analysis:Max. 5 ng/l
- Explosion Limit:36.5 %
:
:74.12 g/mol
:35 °C
:–117,4 °C
:0,713 g/cm³ (20 °C)
:–45 °C
:Ambient
:200-467-2
:1155
:3,I
:Stabilised with ethanol 2%
:GHS02
:GHS07
:60-29-7
:(CH₃CH₂)₂O
:MFCD00011646