Glycine 98.5-101.5% (dried basis), BAKER ANALYZED®, Biochemical Reagent, J.T.Baker®
Supplier: Avantor
Certificates
About this item
CAS Number:56-40-6
:
Formula:NH₂CH₂COOH
MDL Number:MFCD00008131
:75.07 g/mol
Merck Index:13,04504
Boiling Pt:181 °C (1013 hPa)
Melting Pt:83 °C
Density:1,16 g/cm³
Storage Temperature:Ambient
EINECS:200-272-2
Specification Test Results
- Assay (C₂H₅NO₂) (dried basis, by non-aqueous acid- base titration):98.5 - 101.5 %
- Residue after Ignition:≤0.10 %
- Lead (Pb):≤5.00 ppm
- Description:Passes Test
- Solubility:Passes Test
- Loss on Drying at 105°C:≤0.2 %