Ammonium 1-pyrrolidinecarbodithioate ≥98.0%, BAKER ANALYZED®, J.T.Baker®
Catalog # BAKRB337-02
Supplier: Avantor
CAS Number:
Specifications
- Pack type:Glass bottle, amber, wide mouth
- Pk:10 g
- Seq:3236
Specifications
Specification Test Results
- Assay (C₅H₁₂N₂S₂):≥98.0 %
- Insoluble Matter:≤0.10 %
- Melting Point:≥146 °C
- Suitability as Chelating Agent:Passes Test
Specification Test Results
About this item
:
:146.3 · 18.0 g/mol
Melting Pt:≥146 °C
CAS Number:5108-96-3
Formula:C₅H₁₂N₂S₂
MDL Number:MFCD00012720