


Perchloric acid 0.1 N in glacial acetic acid, BAKER ANALYZED® volumetric solution, J.T.Baker®
Catalog # BAKR5624-02
Supplier: Avantor
CAS Number:
Specifications
- Pack type:Glass bottle, amber, narrow mouth
- Pk:1 L
- Seq:5037
Specifications
About this item
:
:100.46 g/mol
Density:1,05 g/cm³
Storage Temperature:Ambient
EINECS:231-512-4
UN:2920
ADR:8, 3,II
:GHS02
:GHS05
:GHS07
CAS Number:7601-90-3
Formula:HClO₄
MDL Number:MFCD00011325