

Diethylenetriamine ≥98%, BAKER ANALYZED®, J.T.Baker®
Catalog # BAKRH768-07
Supplier: Avantor
CAS Number:
Specifications
- Pack type:Glass bottle, amber, narrow mouth
- Pk:500 mL
- Seq:2725
Specifications
Specification Test Results
- Assay:≥98 %
- Refractive Index, n²ºD:1.4815 - 1.4845
Specification Test Results
About this item
:
:103.17 g/mol
Boiling Pt:207 °C (1013 hPa)
Melting Pt:–39 °C
Density:0,96 g/cm³
Flash Pt:102,00 °C
UN:2079
ADR:8,II
:GHS05
:GHS07
Storage Temperature:Ambient
CAS Number:111-40-0
Formula:C₄H₁₃N₃
MDL Number:MFCD00008171