Analytical standard, Canthaxanthin (trans), reference material, Honeywell Fluka™
Catalog # HONCR1364-10MG
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Glass Bottle
- Pk:10 mg
- Standards:Analytical standard, neat
- CAS No.:514-78-3
- Description:Canthaxanthin (trans)
- Melting point:199…205 °C
- Formula:C₄₀H₅₂O₂
- Storage temperature:2…8 °C
- Description:Canthaxanthin (trans)
Specifications
About this item
- High quality organic reference materials produced according to ISO 9001
- Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results