Analytical standard, Dichlorprop-isooctyl, reference material, Honeywell Fluka™
Catalog # HONCR1300-25MG
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Glass Bottle
- Pk:25 mg
- Standards:Analytical standard, neat
- CAS No.:28631-35-8
- Description:Dichlorprop-isooctyl
- Formula:C₁₇H₂₄Cl₂O₃
- Storage temperature:Ambient
- Description:Dichlorprop-isooctyl
Specifications
About this item
- High quality organic reference materials produced according to ISO 9001
- Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results