Analytical standard, Cefquinome sulfate, reference material, Honeywell Fluka™
Catalog # HONCR1259-100MG
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Glass Bottle
- Pk:100 mg
- Standards:Analytical standard, neat
- CAS No.:118443-89-3
- Description:Cefquinome sulfate
- Melting point:190…205 °C
- Formula:C₂₃H₂₄N₆O₅S₂ · H₂SO₄
- Storage temperature:2…8 °C
- Description:Cefquinome sulfate
Specifications
About this item
- High quality organic reference materials produced according to ISO 9001
- Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results