Analytical standard, Desoxycarbadox, reference material, Honeywell Fluka™
Catalog # HONCR1207-10MG
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Glass Bottle
- Pk:10 mg
- Standards:Analytical standard, neat
- CAS No.:55456-55-8
- Description:Desoxycarbadox
- Melting point:245 °C
- Formula:C₁₁H₁₀N₄O₂
- Storage temperature:Ambient
- Description:Desoxycarbadox
Specifications
About this item
- High quality organic reference materials produced according to ISO 9001
- Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results