Analytical standard, Natamycin, reference material, Honeywell Fluka™
Catalog # HONCR1116-50MG
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Glass Bottle
- Pk:50 mg
- Standards:Analytical standard, neat
- CAS No.:7681-93-8
- Description:Natamycin
- Melting point:280 °C
- Formula:C₃₃H₄₇NO₁₃
- Storage temperature:2…8 °C
- Description:Natamycin
Specifications
About this item
- High quality organic reference materials produced according to ISO 9001
- Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results