Order Entry
Singapore
ContactUsLinkComponent
Analytical standard, Ibuprofen, reference material, Honeywell Fluka™
Analytical standard, Ibuprofen, reference material, Honeywell Fluka™
Catalog # HONCR1092-50MG
CAS Number:  
Analytical standard, Ibuprofen, reference material, Honeywell Fluka™
Catalog # HONCR1092-50MG
CAS Number:  

Specifications

  • Pack type:
    Glass Bottle
  • Pk:
    50 mg
  • Standards:
    Analytical standard, neat
  • CAS No.:
    15687-27-1
  • Description:
    Ibuprofen
  • Melting point:
    77…78 °C
  • Formula:
    C₁₃H₁₈O₂
  • Storage temperature:
    Ambient
  • Description:
    Ibuprofen

Specifications

About this item

  • High quality organic reference materials produced according to ISO 9001
  • Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results