Analytical standard, Moxidectin, reference material, Honeywell Fluka™
Catalog # HONCR1065-25MG
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Glass Bottle
- Pk:25 mg
- Standards:Analytical standard, neat
- CAS No.:113507-06-5
- Description:Moxidectin
- Formula:C₃₇H₅₃NO₈
- Storage temperature:–20°C
- Description:Moxidectin
Specifications
About this item
- High quality organic reference materials produced according to ISO 9001
- Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results