
Mineral oil heavy, white stabilised USP, Macron Fine Chemicals™
Supplier: MACRON AVANTOR BRAND
Certificates
About this item
CAS Number:8012-95-1
:
MDL Number:MFCD00131611
Merck Index:13,07263
Density:0,88 g/cm³
:Stabilised with vitamin E 10-20 ppm
EINECS:232-384-2
:GHS08
Specification Test Results
- Bulk Pharmaceutical Chemical:Passes Test
- USP - Acidity:Passes Test
- USP - Limit of Polycyclic Aromatic Hydrocarbons:Passes Test
- USP - Limit of Sulfur Compounds:Passes Test
- USP - Identification A:Passes Test
- USP - Identification B:Passes Test
- USP - Specific Gravity at 25°/25°C:0.845 - 0.905
- USP - Viscosity at 40 C, mm₂/s:34.5 - 150.0
- USP - Readily Carbonizable Substances:Passes Test
- USP - Solid Paraffin:Passes Test