Order Entry
Singapore
ContactUsLinkComponent
Springs for inserts
Springs for inserts
Catalog # MANA702974
Supplier:  MACHEREY-NAGEL
undefined
Springs for inserts
Catalog # MANA702974
Supplier:  MACHEREY-NAGEL
Specifications
  • Description:
    Metal spring for 6×40 mm conical inserts
  • Seq:
    70
  • Pk:
    100